Learn More
Syngene™ Small Wave UV Tube
GREENER_CHOICESDP
Catalog No. 01257294 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Wavelength:
254nm
Catalog No. Wavelength
01-257-294 254nm
1 options
Catalog No. 01-257-294 Supplier Syngene™ Supplier No. SWT8
Add to Cart
Add to Cart
Syngene™ Small Wave UV Tube for use in UV Transilluminators.
  • For use with all GX and GE model transilluminators
  • 254nm wavelength
  • Small wave
Specifications
Intensity Fixed
Wavelength 254nm
Type UV Tube Short Wave
Description Short wave UV tube
For Use With (Equipment) All GX/GE model transilluminators
WARNING: Cancer - www.P65Warnings.ca.gov