Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Thermoplastic, 1/8 in I.D., 1/4 in O.D.
Catalog No. NC2398410
Encompass_Preferred
Saint-Gobain Tygon E-3603 tubing is a crystal-clear, flexible thermoplastic tubing engineered for medical and diagnostic fluid handling where chemical compatibility, visual inspection, and low extractables are required. The tubing features a glassy-smooth inner bore to reduce buildup, and it offers resistance to flex-fatigue and abrasion for consistent fluid transfer in instrumentation.
  • Crystal-clear thermoplastic for visual inspection and low extractables.
  • 1/8 in I.D., 1/4 in O.D., 1/16 in wall, supplied in 100 ft length.
  • Glassy-smooth inner bore reduces buildup and carryover.
  • Resists flex-fatigue and abrasion for extended service life.
  • Suitable for IVD analyzers, diagnostic instrumentation, and general fluid transfer.
  • Maximum recommended operating temperature 165 °F (74 °C); vacuum rating 29.9 inHg (760 mmHg).
Catalog No. NC2398410 Supplier Accuflow Systems Supplier No. ACF02007
Saint-Gobain Tygon E-3603 tubing is a crystal-clear, flexible thermoplastic tubing engineered for medical and diagnostic fluid handling where chemical compatibility, visual inspection, and low extractables are required. The tubing features a glassy-smooth inner bore to reduce buildup, and it offers resistance to flex-fatigue and abrasion for consistent fluid transfer in instrumentation.
  • Crystal-clear thermoplastic for visual inspection and low extractables.
  • 1/8 in I.D., 1/4 in O.D., 1/16 in wall, supplied in 100 ft length.
  • Glassy-smooth inner bore reduces buildup and carryover.
  • Resists flex-fatigue and abrasion for extended service life.
  • Suitable for IVD analyzers, diagnostic instrumentation, and general fluid transfer.
  • Maximum recommended operating temperature 165 °F (74 °C); vacuum rating 29.9 inHg (760 mmHg).