Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 1/8 in ID, 1/4 in OD, Clear
Catalog No. NC2386966
Encompass_Preferred
Saint-Gobain Tygon B-44-4X tubing is a clear, food-contact flexible tubing engineered for dairy, beverage, and dispensing applications. It features a smooth, non-porous bore that resists particulate entrapment and is compatible with common sanitary cleaners, providing reliable fluid transfer in food-processing systems.
  • Clear, smooth, non-porous bore that resists particulate entrapment.
  • Phthalate- and BPA-free formulation suitable for food contact.
  • Inner diameter 1/8 in (3.18 mm), outer diameter 1/4 in (6.35 mm).
  • Maximum working pressure 60 psi (4.1 bar) at 73°F (23°C).
  • Maximum recommended operating temperature 165°F (74°C).
  • Minimum bend radius 3/8 in (9.53 mm).
  • Complies with FDA food additive regulations, NSF-51, NSF-61, 3-A, REACH, and EU food-contact regulations.
  • Resistant to harsh alkaline cleaners and sanitizers.
Catalog No. NC2386966 Supplier Accuflow Systems Supplier No. AAA00007
Saint-Gobain Tygon B-44-4X tubing is a clear, food-contact flexible tubing engineered for dairy, beverage, and dispensing applications. It features a smooth, non-porous bore that resists particulate entrapment and is compatible with common sanitary cleaners, providing reliable fluid transfer in food-processing systems.
  • Clear, smooth, non-porous bore that resists particulate entrapment.
  • Phthalate- and BPA-free formulation suitable for food contact.
  • Inner diameter 1/8 in (3.18 mm), outer diameter 1/4 in (6.35 mm).
  • Maximum working pressure 60 psi (4.1 bar) at 73°F (23°C).
  • Maximum recommended operating temperature 165°F (74°C).
  • Minimum bend radius 3/8 in (9.53 mm).
  • Complies with FDA food additive regulations, NSF-51, NSF-61, 3-A, REACH, and EU food-contact regulations.
  • Resistant to harsh alkaline cleaners and sanitizers.