Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Non-DEHP, 1/16 in I.D., 50 ft
Catalog No. 502365776
Encompass_Preferred
Clear, flexible laboratory tubing formulated with a bio-based plasticizer for resistance to flex-fatigue and abrasion. The non-DEHP formulation offers low extractables and is suitable for analytical, biopharmaceutical, and instrumentation connections. Supplied in 50 ft coils for convenient bench and instrument use.
  • Clear, flexible PVC tubing.
  • 1/16 in (1.6 mm) inner diameter.
  • 3/16 in (4.8 mm) outer diameter.
  • 1/16 in (1.6 mm) wall thickness.
  • 50 ft (15 m) package length.
  • Formulation resists flex-fatigue and abrasion.
  • Non-DEHP, bio-based plasticizer for reduced extractables.
  • Typical use: laboratory connections, analytical instrumentation, gas and drain lines.
Catalog No. 50-236-5776 Supplier Accuflow Systems Supplier No. ACF00003
Clear, flexible laboratory tubing formulated with a bio-based plasticizer for resistance to flex-fatigue and abrasion. The non-DEHP formulation offers low extractables and is suitable for analytical, biopharmaceutical, and instrumentation connections. Supplied in 50 ft coils for convenient bench and instrument use.
  • Clear, flexible PVC tubing.
  • 1/16 in (1.6 mm) inner diameter.
  • 3/16 in (4.8 mm) outer diameter.
  • 1/16 in (1.6 mm) wall thickness.
  • 50 ft (15 m) package length.
  • Formulation resists flex-fatigue and abrasion.
  • Non-DEHP, bio-based plasticizer for reduced extractables.
  • Typical use: laboratory connections, analytical instrumentation, gas and drain lines.