Learn More
Accuflow Systems Saint-Gobain Tygon S3 Tubing, 1/4 in ID, 3/8 in OD
Catalog No. 502365821
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 tubing is a clear, flexible thermoplastic elastomer designed for laboratory and vacuum applications. It uses a non-DEHP bio-based plasticizer for improved flex-fatigue resistance and environmental profile, and provides a smooth inner bore and chemical resistance for reliable fluid transfer in peristaltic pumps and general laboratory connections.
  • Inside diameter: 1/4 in.
  • Outside diameter: 3/8 in.
  • Wall thickness: 1/16 in.
  • Package length: 100 ft.
  • Material: thermoplastic elastomer (non-DEHP).
  • Chemical resistant and non-oxidizing.
  • Smooth, polished inner bore for consistent flow.
  • Resistant to flex-fatigue and abrasion; suitable for peristaltic pumps.
Catalog No. 50-236-5821 Supplier Accuflow Systems Supplier No. ACF02017
Saint-Gobain Tygon S3 E-3603 tubing is a clear, flexible thermoplastic elastomer designed for laboratory and vacuum applications. It uses a non-DEHP bio-based plasticizer for improved flex-fatigue resistance and environmental profile, and provides a smooth inner bore and chemical resistance for reliable fluid transfer in peristaltic pumps and general laboratory connections.
  • Inside diameter: 1/4 in.
  • Outside diameter: 3/8 in.
  • Wall thickness: 1/16 in.
  • Package length: 100 ft.
  • Material: thermoplastic elastomer (non-DEHP).
  • Chemical resistant and non-oxidizing.
  • Smooth, polished inner bore for consistent flow.
  • Resistant to flex-fatigue and abrasion; suitable for peristaltic pumps.