Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Non-DEHP, 1/4 in ID, 3/8 in OD
Catalog No. 502365886
Encompass_Preferred
Tygon S3 E-3603 is a non-DEHP laboratory tubing formulated for chemical compatibility, visual clarity, and durable performance in analytical and fluid-transfer applications. It offers a glassy-smooth inner bore to reduce carryover and resists flex-fatigue and abrasion.
  • Non-DEHP PVC formulation with bio-based plasticizer; no BPA or phthalates intentionally added.
  • 1/4 in (6.4 mm) I.D. x 3/8 in (9.5 mm) O.D., 1/16 in (1.6 mm) wall.
  • Available in 500 ft (152 m) coil length.
  • Crystal-clear, glassy-smooth inner bore for visual inspection and reduced carryover.
  • High dimensional tolerance, resistant to flex-fatigue and abrasion.
  • Suitable for laboratory connections, analytical instrumentation, vacuum pumps, and general fluid lines.
Catalog No. 50-236-5886 Supplier Accuflow Systems Supplier No. ACF04017
Tygon S3 E-3603 is a non-DEHP laboratory tubing formulated for chemical compatibility, visual clarity, and durable performance in analytical and fluid-transfer applications. It offers a glassy-smooth inner bore to reduce carryover and resists flex-fatigue and abrasion.
  • Non-DEHP PVC formulation with bio-based plasticizer; no BPA or phthalates intentionally added.
  • 1/4 in (6.4 mm) I.D. x 3/8 in (9.5 mm) O.D., 1/16 in (1.6 mm) wall.
  • Available in 500 ft (152 m) coil length.
  • Crystal-clear, glassy-smooth inner bore for visual inspection and reduced carryover.
  • High dimensional tolerance, resistant to flex-fatigue and abrasion.
  • Suitable for laboratory connections, analytical instrumentation, vacuum pumps, and general fluid lines.