Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, PVC, 1/4 in ID, 3/8 in OD
Catalog No. 502365860
Encompass_Preferred
Tygon S3 B-44-3 is a clear, flexible beverage-grade tubing engineered for food and beverage transfer and dispensing. It resists taste and odor transfer, offers excellent non-wetting properties for complete drainage, and is compatible with common sanitizing agents.
  • Clear, flexible PVC construction with no BPA or phthalates.
  • 1/4 in inside diameter, 3/8 in outside diameter, 1/16 in wall thickness.
  • Typically supplied on 50 ft rolls for easy installation.
  • Maximum working pressure 34 psi at 73°F (2.3 bar at 23°C).
  • Shore A hardness 66 for flexible, kink-resistant performance.
  • Glass-like clarity allows visual flow monitoring.
  • Complies with FDA food additive regulations, NSF-51, NSF-61, 3-A, EU 1935/2004, and REACH.
Catalog No. 50-236-5860 Supplier Accuflow Systems Supplier No. AAB00017
Tygon S3 B-44-3 is a clear, flexible beverage-grade tubing engineered for food and beverage transfer and dispensing. It resists taste and odor transfer, offers excellent non-wetting properties for complete drainage, and is compatible with common sanitizing agents.
  • Clear, flexible PVC construction with no BPA or phthalates.
  • 1/4 in inside diameter, 3/8 in outside diameter, 1/16 in wall thickness.
  • Typically supplied on 50 ft rolls for easy installation.
  • Maximum working pressure 34 psi at 73°F (2.3 bar at 23°C).
  • Shore A hardness 66 for flexible, kink-resistant performance.
  • Glass-like clarity allows visual flow monitoring.
  • Complies with FDA food additive regulations, NSF-51, NSF-61, 3-A, EU 1935/2004, and REACH.