Learn More
Accuflow Systems Saint-Gobain Tygon Laboratory Tubing, Phthalate-Free, 1/4 in I.D., 50 ft
Catalog No. 502365865
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 is a phthalate-free, bio-based, clear flexible laboratory tubing formulated for chemical compatibility, visual inspection, and vacuum-capable fluid transfer. Typical sold configuration for the referenced SKU is 1/4 in inside diameter (≈6.35 mm), 5/16 in outside diameter (≈7.94 mm), 0.03 in wall (≈0.79 mm), supplied in 50 ft coils. The formulation provides a smooth inner bore, lot-to-lot consistency, and regulatory compliance for laboratory and food-contact applications.
  • Phthalate-free, bio-based plasticizer formulation.
  • Smooth, polished inner wall to help prevent buildup.
  • Suitable for vacuum and peristaltic pump applications.
  • Non-toxic and non-contaminating; contains no BPA or phthalates.
  • Meets USP Class VI, (EU) 10/2011, (EC) 1935/2004, and FDA food-contact criteria.
  • Fully autoclavable and sterilizable.
  • Standard sizes available to hold full vacuum at room temperature.
Catalog No. 50-236-5865 Supplier Accuflow Systems Supplier No. ACF00016
Saint-Gobain Tygon S3 E-3603 is a phthalate-free, bio-based, clear flexible laboratory tubing formulated for chemical compatibility, visual inspection, and vacuum-capable fluid transfer. Typical sold configuration for the referenced SKU is 1/4 in inside diameter (≈6.35 mm), 5/16 in outside diameter (≈7.94 mm), 0.03 in wall (≈0.79 mm), supplied in 50 ft coils. The formulation provides a smooth inner bore, lot-to-lot consistency, and regulatory compliance for laboratory and food-contact applications.
  • Phthalate-free, bio-based plasticizer formulation.
  • Smooth, polished inner wall to help prevent buildup.
  • Suitable for vacuum and peristaltic pump applications.
  • Non-toxic and non-contaminating; contains no BPA or phthalates.
  • Meets USP Class VI, (EU) 10/2011, (EC) 1935/2004, and FDA food-contact criteria.
  • Fully autoclavable and sterilizable.
  • Standard sizes available to hold full vacuum at room temperature.