Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 1/8 in x 1/4 in, 500 ft
Catalog No. 502365820
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 laboratory tubing is a clear, flexible PVC tubing formulated for instrumentation connection, venting, draining, and peristaltic pump use. It uses a non-DEHP, bio-based plasticizer and contains no phthalates or BPA, providing chemical resistance, abrasion resistance, and extended service life.
  • 1/8 in inner diameter, 1/4 in outer diameter, 500 ft length
  • Wall thickness approximately 1/16 in
  • Non-DEHP bio-based plasticizer; phthalate-free and BPA-free
  • Crystal clear PVC with smooth inner wall for reproducible results
  • Suitable for instrumentation connection, vent, drain, and peristaltic pump applications
Catalog No. 50-236-5820 Supplier Accuflow Systems Supplier No. ACF04007
Saint-Gobain Tygon S3 E-3603 laboratory tubing is a clear, flexible PVC tubing formulated for instrumentation connection, venting, draining, and peristaltic pump use. It uses a non-DEHP, bio-based plasticizer and contains no phthalates or BPA, providing chemical resistance, abrasion resistance, and extended service life.
  • 1/8 in inner diameter, 1/4 in outer diameter, 500 ft length
  • Wall thickness approximately 1/16 in
  • Non-DEHP bio-based plasticizer; phthalate-free and BPA-free
  • Crystal clear PVC with smooth inner wall for reproducible results
  • Suitable for instrumentation connection, vent, drain, and peristaltic pump applications