Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Beverage, 1/8 in ID, 1/4 in OD
Catalog No. 502365898
Encompass_Preferred
Saint-Gobain Tygon B-44-3 beverage tubing is a clear, flexible thermoplastic elastomer tube formulated for food and beverage transfer and dispensing. It is phthalate-free, non-wetting for complete drainage and cleaning, and offers glass-like clarity for visual flow monitoring.
  • Beverage-grade thermoplastic elastomer construction, phthalate-free.
  • 1/8 in (3.18 mm) inner diameter, 1/4 in (6.35 mm) outer diameter, 1/16 in (1.59 mm) wall.
  • 50 ft coil packaging for bulk fluid handling.
  • Compliant with NSF-51, NSF-61, FDA, EU food contact, and Japan Food Sanitation Law.
  • Non-wetting surface for easy draining and cleaning, with glass-like clarity for visual inspection.
Catalog No. 50-236-5898 Supplier Accuflow Systems Supplier No. AAB00007
Saint-Gobain Tygon B-44-3 beverage tubing is a clear, flexible thermoplastic elastomer tube formulated for food and beverage transfer and dispensing. It is phthalate-free, non-wetting for complete drainage and cleaning, and offers glass-like clarity for visual flow monitoring.
  • Beverage-grade thermoplastic elastomer construction, phthalate-free.
  • 1/8 in (3.18 mm) inner diameter, 1/4 in (6.35 mm) outer diameter, 1/16 in (1.59 mm) wall.
  • 50 ft coil packaging for bulk fluid handling.
  • Compliant with NSF-51, NSF-61, FDA, EU food contact, and Japan Food Sanitation Law.
  • Non-wetting surface for easy draining and cleaning, with glass-like clarity for visual inspection.