Learn More
Accuflow Systems Saint-Gobain Tubing, Tygon E-3603, 3/16 in ID, 50 ft
Catalog No. 502365763
Encompass_Preferred
Tygon E-3603 tubing is a clear, flexible laboratory PVC tubing designed for chemical resistance, low contamination, and extended service life in instrumentation and general laboratory fluid handling. It features a glassy-smooth inner bore to reduce buildup and is free of BPA and phthalates.
  • 3/16 in (4.8 mm) inner diameter, 3/8 in (9.5 mm) outer diameter, 3/32 in (2.4 mm) wall, 50 ft (15.2 m) length.
  • PVC construction provides chemical resistance and flexibility.
  • Contains no BPA or phthalates; non-oxidizing and non-contaminating.
  • Maximum working pressure 36 psi (2.5 bar) at 73°F (23°C); vacuum rating 29.9 inHg.
  • Glassy-smooth inner bore reduces buildup and facilitates cleaning.
  • Suitable for laboratory instrumentation, peristaltic and vacuum pumps, condensers, and gas or drain lines.
Catalog No. 50-236-5763 Supplier Accuflow Systems Supplier No. ACF00013
Tygon E-3603 tubing is a clear, flexible laboratory PVC tubing designed for chemical resistance, low contamination, and extended service life in instrumentation and general laboratory fluid handling. It features a glassy-smooth inner bore to reduce buildup and is free of BPA and phthalates.
  • 3/16 in (4.8 mm) inner diameter, 3/8 in (9.5 mm) outer diameter, 3/32 in (2.4 mm) wall, 50 ft (15.2 m) length.
  • PVC construction provides chemical resistance and flexibility.
  • Contains no BPA or phthalates; non-oxidizing and non-contaminating.
  • Maximum working pressure 36 psi (2.5 bar) at 73°F (23°C); vacuum rating 29.9 inHg.
  • Glassy-smooth inner bore reduces buildup and facilitates cleaning.
  • Suitable for laboratory instrumentation, peristaltic and vacuum pumps, condensers, and gas or drain lines.