Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 3/32 x 5/32 in, 50 ft
Catalog No. 502365843
Encompass_Preferred
Saint-Gobain Tygon B-44-3 beverage tubing is a clear, flexible tube formulated for food and beverage transfer and dispensing. It provides non-wetting properties for complete drainage, resists common sanitizing solutions, and meets major food-contact regulations for beverage applications.
  • Clear, flexible tubing for beverage transfer and dispensing.
  • 3/32 in ID x 5/32 in OD, 50 ft length.
  • Wall thickness 1/32 in for consistent flow and fit.
  • Shore A 66 hardness for balance of flexibility and durability.
  • Maximum recommended operating temperature 165°F (74°C).
  • Complies with FDA, NSF-51, NSF-61, 3-A, REACH, and EU 10/2011 where applicable.
  • Contains no BPA or phthalates; not listed under California Proposition 65.
  • Excellent resistance to sanitizing solutions and easy to flush clean.
Catalog No. 50-236-5843 Supplier Accuflow Systems Supplier No. AAB00004
Saint-Gobain Tygon B-44-3 beverage tubing is a clear, flexible tube formulated for food and beverage transfer and dispensing. It provides non-wetting properties for complete drainage, resists common sanitizing solutions, and meets major food-contact regulations for beverage applications.
  • Clear, flexible tubing for beverage transfer and dispensing.
  • 3/32 in ID x 5/32 in OD, 50 ft length.
  • Wall thickness 1/32 in for consistent flow and fit.
  • Shore A 66 hardness for balance of flexibility and durability.
  • Maximum recommended operating temperature 165°F (74°C).
  • Complies with FDA, NSF-51, NSF-61, 3-A, REACH, and EU 10/2011 where applicable.
  • Contains no BPA or phthalates; not listed under California Proposition 65.
  • Excellent resistance to sanitizing solutions and easy to flush clean.