Learn More
Accuflow Systems Saint-Gobain Tygon E-3603 Tubing, Non-DEHP, 3/4 in I.D., 50 ft
Catalog No. 502365926
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 tubing is a flexible, crystal-clear, non-DEHP thermoplastic tubing formulated for laboratory and analytical fluid transfer. It provides improved resistance to flex-fatigue and abrasion for longer service life in peristaltic pump and vacuum applications, while maintaining good chemical compatibility and low extractables for instrument and food-contact uses.
  • Crystal clear for visual inspection and monitoring.
  • Resistant to flex-fatigue and abrasion for extended life.
  • Smooth inner bore reduces buildup and facilitates cleaning.
  • Suitable for peristaltic pumps, vacuum lines, and analytical instrumentation.
  • Dimensions: 3/4 in I.D., 1-1/4 in O.D., 1/4 in wall, 50 ft (15 m) length.
Catalog No. 50-236-5926 Supplier Accuflow Systems Supplier No. ACF00057
Saint-Gobain Tygon S3 E-3603 tubing is a flexible, crystal-clear, non-DEHP thermoplastic tubing formulated for laboratory and analytical fluid transfer. It provides improved resistance to flex-fatigue and abrasion for longer service life in peristaltic pump and vacuum applications, while maintaining good chemical compatibility and low extractables for instrument and food-contact uses.
  • Crystal clear for visual inspection and monitoring.
  • Resistant to flex-fatigue and abrasion for extended life.
  • Smooth inner bore reduces buildup and facilitates cleaning.
  • Suitable for peristaltic pumps, vacuum lines, and analytical instrumentation.
  • Dimensions: 3/4 in I.D., 1-1/4 in O.D., 1/4 in wall, 50 ft (15 m) length.