Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 3/8 in ID, 5/8 in OD, Clear
Catalog No. 502365897
Encompass_Preferred
Tygon B-44-3 beverage tubing is a clear, flexible tubing formulated for food and beverage transfer. It resists flavor and odor transfer, offers excellent chemical resistance to common sanitizers, and provides reliable performance across a wide temperature range.
  • Food- and beverage-grade formulation, conforms to FDA and NSF standards.
  • Clear, glass-like clarity for visual monitoring of flow.
  • 3/8 in inside diameter, 5/8 in outside diameter, 0.13 in wall thickness.
  • 50 ft roll length.
  • Maximum recommended operating temperature 165 °F (74 °C); cold flexibility to -67 °F (-55 °C).
  • Minimum bend radius 1-1/8 in; maximum working pressure 44 psi at 73 °F.
Catalog No. 50-236-5897 Supplier Accuflow Systems Supplier No. AAB00029
Tygon B-44-3 beverage tubing is a clear, flexible tubing formulated for food and beverage transfer. It resists flavor and odor transfer, offers excellent chemical resistance to common sanitizers, and provides reliable performance across a wide temperature range.
  • Food- and beverage-grade formulation, conforms to FDA and NSF standards.
  • Clear, glass-like clarity for visual monitoring of flow.
  • 3/8 in inside diameter, 5/8 in outside diameter, 0.13 in wall thickness.
  • 50 ft roll length.
  • Maximum recommended operating temperature 165 °F (74 °C); cold flexibility to -67 °F (-55 °C).
  • Minimum bend radius 1-1/8 in; maximum working pressure 44 psi at 73 °F.