Saint-Gobain Tygon S3 E-3603 tubing is a clear, flexible thermoplastic elastomer designed for laboratory and vacuum fluid transfer applications. The formulation resists flex-fatigue and abrasion, features a glassy smooth inner bore to reduce buildup, and contains no intentionally added phthalates or BPA. This size is 5/32 in ID x 9/32 in OD with a 1/16 in wall and is supplied in 50 ft lengths marked at 1 ft intervals for easy measuring.
- Crystal clear thermoplastic elastomer construction.
- 5/32 in internal diameter x 9/32 in outside diameter, 1/16 in wall.
- 50 ft package length, marked at 1 ft intervals for measuring.
- Withstands full vacuum at room temperature (29.9 in Hg).
- Maximum recommended operating temperature 165°F (74°C).
- Resistant to flex-fatigue and abrasion for extended service life.
- Phthalate-free formulation; no BPA intentionally added.