Learn More
Beckman Coulter Microfuge Tubes, 0.5mL, Graduated, Polypropylene, Natural, Bag of 1000
Catalog No. NC9805566
Encompass_Preferred
The Beckman Coulter Ultra-Clear Centrifuge Tubes are 0.5mL polypropylene conical tubes designed for high speed centrifugation. They feature a graduated scale in mL to allow accurate volume measurement and an extra-thick tube wall for increased strength and rigidity. The tubes have a flat top and uniform tip shape to ensure consistent pellet formation. Their clear polypropylene construction allows easy sample visibility. The tubes can withstand RCF up to 30,000 x g and autoclaving at 121°C for 20 minutes. They come sterile and individually wrapped in packs of 500.
Catalog No. NC9805566 Supplier Beckman Coulter Supplier No. 344319
The Beckman Coulter Ultra-Clear Centrifuge Tubes are 0.5mL polypropylene conical tubes designed for high speed centrifugation. They feature a graduated scale in mL to allow accurate volume measurement and an extra-thick tube wall for increased strength and rigidity. The tubes have a flat top and uniform tip shape to ensure consistent pellet formation. Their clear polypropylene construction allows easy sample visibility. The tubes can withstand RCF up to 30,000 x g and autoclaving at 121°C for 20 minutes. They come sterile and individually wrapped in packs of 500.