Learn More
Cole-Parmer Flow Through Tubing Weight, PTFE, 3/16" to 1/4" Tubing
Supplier:  Cole-Parmer 5011798
Encompass_Preferred
Cole-Parmer flow-through tubing weights provide the perfect solution for keeping tubing submerged, preventing floatation, and ensuring uninterrupted flow throughout the fluid transfer process. Crafted from high-quality PTFE, these tubing weights are chemically inert, ensuring compatibility with a wide range of fluids and preventing contamination. Weighted ends are open in the center and feature a tapered hose barb fitting for easy attachment to flexible tubing , ensuring a snug and secure fit. Designed for maximum convenience and versatility, our tubing weights are autoclavable at 121°C for 20 minutes at 15 psi.
Catalog No. 50-255-7790