Learn More
Enterprise Technology Solutions Welch Allyn OAE Hearing Screener Probe Tube (#39421 / 100 per Box)
SDP
Catalog No. 502407024
Encompass_Preferred
The Welch Allyn OAE Hearing Screener Probe Tube (#39421 / 100 per Box) is designed for use with the OAE Hearing Screener (#39500). A clear probe tube must be attached to the probe head before a disposable MicroTymp 4 Ear Tip or Single Use OAE Ear Tip is applied.
Catalog No. 50-240-7024 Supplier Enterprise Technology Solutions Supplier No. WLA39421
The Welch Allyn OAE Hearing Screener Probe Tube (#39421 / 100 per Box) is designed for use with the OAE Hearing Screener (#39500). A clear probe tube must be attached to the probe head before a disposable MicroTymp 4 Ear Tip or Single Use OAE Ear Tip is applied.