Learn More
Research Products International Corp Tube Cube Test Tube Support System, Standard Colors
SDP
Supplier:  Research Products International Corp 140441
Encompass
Multi-option test tube support system accommodates a variety of tube sizes and styles. Each cube can be rotated independently within the tray for a particular tube size. Tube Cube modules and trays are constructed of durable polypropylene and can be autoclaved. Cube modules are color coded for convenience in sample identification.
Each Tube Cube set consists of four removable modules and one storage tray.
Catalog No. 50-550-159