Learn More
Med Vet International VACUETTE Venous Blood Collection Tube Lithium Heparin / Separator Gel Additive 5 mL Pull Cap Polyethylene Terephthalate (PET) Tube, 456087P, 1200/Case
Supplier:  Med Vet International 416954CS
Encompass_Preferred
The interior of these analyte determination tubes is coated with lithium heparin – an anticoagulant that prevents blood from clotting by inhibiting the action of clotting factors Green pull cap and yellow ring Dimensions: 13 x 100 millimeters Volume is 5 mL Sterile
Catalog No. 50-304-4800