Learn More
Med Vet International BD Vacutainer Venous Blood Collection Tube Plain 10 mL Conventional Closure Glass Tube, 366430, 100/Box
Catalog No. 503044047
Encompass_Preferred
Manufacturer #366430 Brand BD Vacutainer Manufacturer BD Country of Origin United States Additive Plain Application Venous Blood Collection Tube Closure Color Red Closure Type Conventional Closure Coating Silicone Coated Tube Color Code Red Dimensions 16 X 100 mm
Catalog No. 50-304-4047 Supplier Med Vet International Supplier No. 1159BX
Manufacturer #366430 Brand BD Vacutainer Manufacturer BD Country of Origin United States Additive Plain Application Venous Blood Collection Tube Closure Color Red Closure Type Conventional Closure Coating Silicone Coated Tube Color Code Red Dimensions 16 X 100 mm