Learn More
Selleck Chemical LLC Tubeimoside I S3814-10mg
SDP
Supplier:  Selleck Chemical LLC S381410MG
Encompass_Preferred
Tubeimoside I (Lobatoside H TBMS1) a triterpenoid saponin isolated from the tubers of Bolbostemma paniculatum shows potent antitumor and antitumor-promoting effects
Catalog No. 50-310-9804