Learn More
Medchemexpress LLC Tubeimoside III | 115810-13-4 | 1 ML
SDP
Catalog No. 5000421149
Encompass_Preferred
Tubeimoside III is a triterpenoid saponin with anti-inflammatory and antitumor effects. It has an LD50 of 15 mg/kg by intraperitoneal injection in ICR mice. It is used for the research of tumors and inflammatory diseases. It is for research use only and not sold to patients. The product has a purity of 99.91%.
  • Triterpenoid saponin
  • Anti-inflammatory effects
  • Antitumor effects
  • Used for the research of tumors and inflammatory diseases
  • Inhibits NO production and reduces inflammatory factors in LPS-induced RAW264.7 cells
  • Exhibits anti-inflammatory activity in an LPS-induced acute inflammation mouse model
  • Shows anti-inflammatory activity by inhibiting ear edema in a TPA-induced mouse ear edema model
  • Possesses antitumor activity in mice inoculated with S180 sarcoma cells
Catalog No. 50-004-21149 Supplier Medchemexpress LLC Supplier No. HYN254210MM/1ML
Tubeimoside III is a triterpenoid saponin with anti-inflammatory and antitumor effects. It has an LD50 of 15 mg/kg by intraperitoneal injection in ICR mice. It is used for the research of tumors and inflammatory diseases. It is for research use only and not sold to patients. The product has a purity of 99.91%.
  • Triterpenoid saponin
  • Anti-inflammatory effects
  • Antitumor effects
  • Used for the research of tumors and inflammatory diseases
  • Inhibits NO production and reduces inflammatory factors in LPS-induced RAW264.7 cells
  • Exhibits anti-inflammatory activity in an LPS-induced acute inflammation mouse model
  • Shows anti-inflammatory activity by inhibiting ear edema in a TPA-induced mouse ear edema model
  • Possesses antitumor activity in mice inoculated with S180 sarcoma cells