Learn More
Perkin Elmer US LLC Flared PVC Two-Stop Peristaltic Pump Tubing - 0.25 mm I.D., Orange/Blue, Pkg. 12
Catalog No. 509052808
Encompass_Preferred
This flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.
Catalog No. 50-905-2808 Supplier Perkin Elmer US LLC Supplier No. N0773112
This flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.