Learn More
TARGETMOL CHEMICALS INC TUBERCIDIN 25MG
SDP
Catalog No. 502576368
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tubercidin (Sparsomycin A) an adenosine analogue is a nucleoside antibiotic. It is incorporated into DNA and inhibits polymerases thereby inhibiting DNA replication and RNA and protein synthesis. This agent also exhibits antifungal and antiviral activities. purity: 99%
Catalog No. 50-257-6368 Supplier TARGETMOL CHEMICALS INC Supplier No. T700425MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tubercidin (Sparsomycin A) an adenosine analogue is a nucleoside antibiotic. It is incorporated into DNA and inhibits polymerases thereby inhibiting DNA replication and RNA and protein synthesis. This agent also exhibits antifungal and antiviral activities. purity: 99%