Learn More
Medchemexpress LLC HY-100126 5mg Medchemexpress, Tubercidin CAS:69-33-0 Purity:>98%
SDP
Catalog No. 501931722
Encompass_Preferred
Medchemexpress, HY-100126 5mg Tubercidin CAS:69-33-0 Tubercidin (7-Deazaadenosine) is an antibiotic obtained from Streptomyces tubercidicus, it inhibits the growth of Streptococcus faecalis (8043) by IC 50 of 0.02 μM [1] . Tubercidin (7-Deazaadenosine) inhibits polymerases by incorporating DNA or RNA , thereby inhibiting DNA replication, RNA and protein synthesis [2] . Tubercidin (7-Deazaadenosine) is a weak inhibitor of adenosine phosphorylase, and interferes with the phosphorylation of adenosine and AMP [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1722 Supplier Medchemexpress LLC Supplier No. HY1001265MG
Medchemexpress, HY-100126 5mg Tubercidin CAS:69-33-0 Tubercidin (7-Deazaadenosine) is an antibiotic obtained from Streptomyces tubercidicus, it inhibits the growth of Streptococcus faecalis (8043) by IC 50 of 0.02 μM [1] . Tubercidin (7-Deazaadenosine) inhibits polymerases by incorporating DNA or RNA , thereby inhibiting DNA replication, RNA and protein synthesis [2] . Tubercidin (7-Deazaadenosine) is a weak inhibitor of adenosine phosphorylase, and interferes with the phosphorylation of adenosine and AMP [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.