Learn More
American Radiolabeled Chemicals Inc TUBERCIDINE 3H 1LC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 18161MC
Encompass
Tubercidin-8-3H 1mCi in ethanol:water (1:1), 1 mCi/ml, S.A. 10-20 Ci/mmol, M.W. 266.25
Catalog No. 50-901-233
Add to Cart
Request a Quote