Learn More
Medchemexpress LLC Tuberculosis inhibitor 3 | 2219325-28-5 | 98.5% | 524.48 | 10 MG
SDP
Catalog No. 5000436295
Encompass_Preferred
Tuberculosis inhibitor 3 (compound 2i) displays potent anti-TB activity (MIC < 0.016 μg/mL) against agent-sensitive/resistant MTB strains. It shows acceptable PK profiles with oral bioavailability. For research use only. We do not sell to patients.
  • Exhibits potent anti-TB activity (MIC < 0.016 μg/mL) against agent-sensitive/resistant MTB strains.
  • Shows acceptable PK profiles with oral bioavailability.
  • Demonstrates acceptable PK properties with an AUC of 2489 h ng/mL and a Tmax of 0.25 h.
Catalog No. 50-004-36295 Supplier Medchemexpress LLC Supplier No. HY11414710MG
Tuberculosis inhibitor 3 (compound 2i) displays potent anti-TB activity (MIC < 0.016 μg/mL) against agent-sensitive/resistant MTB strains. It shows acceptable PK profiles with oral bioavailability. For research use only. We do not sell to patients.
  • Exhibits potent anti-TB activity (MIC < 0.016 μg/mL) against agent-sensitive/resistant MTB strains.
  • Shows acceptable PK profiles with oral bioavailability.
  • Demonstrates acceptable PK properties with an AUC of 2489 h ng/mL and a Tmax of 0.25 h.