Learn More
Medchemexpress LLC HY-114147 5mg Medchemexpress, Tuberculosis inhibitor 3 CAS:2219325-28-5 Purity:>98%
SDP
Catalog No. 502080867
Encompass_Preferred
Medchemexpress, HY-114147 5mg Tuberculosis inhibitor 3 CAS:2219325-28-5 Tuberculosis inhibitor 3 (compound 2i) displays potent anti-TB activity (MIC < 0.016 µg/mL) against drug-sensitive/resistant MTB strains. Tuberculosis inhibitor 3 (compound 2i) shows acceptable PK profiles with oral bioavailability. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0867 Supplier Medchemexpress LLC Supplier No. HY1141475MG
Medchemexpress, HY-114147 5mg Tuberculosis inhibitor 3 CAS:2219325-28-5 Tuberculosis inhibitor 3 (compound 2i) displays potent anti-TB activity (MIC < 0.016 µg/mL) against drug-sensitive/resistant MTB strains. Tuberculosis inhibitor 3 (compound 2i) shows acceptable PK profiles with oral bioavailability. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.