Learn More
LabScientific HDPE Coated Wire Tube Racks fit most 30-33mm bottles, vials, tubes. 6 x 2 format. 12 place. Suitable for universal, McCartney and Scintillation vials. Fits 25mL conical Eppendorf vials.
Catalog No. NC2053757
Encompass
HDPE Coated Wire Tube Racks fit most 30-33mm bottles, vials, tubes. 12 place racks feature a chemically-resistant HDPE coated wire and can sustain temperatures to -40 C.Suitable for conical, round or flat bottom, glass or platsic, tubes or bottles. The item is also suitable for universal McCartney and Scintillation vials as well as low-profile tubes up to 33 dia. It features a mesh bottom for support and is autoclavable.
Catalog No. NC2053757 Supplier LabScientific Supplier No. TR3312
HDPE Coated Wire Tube Racks fit most 30-33mm bottles, vials, tubes. 12 place racks feature a chemically-resistant HDPE coated wire and can sustain temperatures to -40 C.Suitable for conical, round or flat bottom, glass or platsic, tubes or bottles. The item is also suitable for universal McCartney and Scintillation vials as well as low-profile tubes up to 33 dia. It features a mesh bottom for support and is autoclavable.