Learn More
LabScientific HDPE COATED WIRE RACK FITS BOTTLES 55-58MM. Ideal for Urine Specimen Containers up to 58mm diameter. Durable racks for transport or sample handling. Slotted bottom. Mesh bottom support. Autoclavable.
Catalog No. NC2053756
Encompass
Durable, coated, racks for transport or sample handling. Ideal for Urine sampling. This item features a slotted bottom, a mesh bottom for suport, and is autoclavable.
Catalog No. NC2053756 Supplier LabScientific Supplier No. TR5824
Durable, coated, racks for transport or sample handling. Ideal for Urine sampling. This item features a slotted bottom, a mesh bottom for suport, and is autoclavable.