Learn More
Cayman Chemical TubermycIn B 10mg
SDP
Catalog No. 502947847
Encompass_Preferred
A bacterial metabolite; active against A. flavus, C. albicans, F. oxysporum, R. solani, and T. rubrum (MICs = 64, 8, 125, 32, and 4 UG/ml, respectively); prevents the development of foliar anthracnose lesions in C. orbiculare-infected cucumber plants
Catalog No. 50-294-7847 Supplier Cayman Chemical Supplier No. 3365910 MG
A bacterial metabolite; active against A. flavus, C. albicans, F. oxysporum, R. solani, and T. rubrum (MICs = 64, 8, 125, 32, and 4 UG/ml, respectively); prevents the development of foliar anthracnose lesions in C. orbiculare-infected cucumber plants