Learn More
Cayman Chemical TuberostemonIn 10mg
SDP
Catalog No. 502947982
Encompass_Preferred
An alkaloid; exhibits feeding deterrent and repellant activities against S. littoralis fifth instar larvae when applied to lettuce leaf disks at 0.01 and 0.1 UG/cm2, respectively; reduces the number of citric acid-induced coughs in guinea pigs at 100 mg/kg; decreases BALF neutrophil and macrophage infiltration and reduces peribronchial and perivascular inflammatory cell infiltration in a cigarette smoke-induced mouse model of acute lung inflammation from 1-10 mg/kg
Catalog No. 50-294-7982 Supplier Cayman Chemical Supplier No. 3445710 MG
An alkaloid; exhibits feeding deterrent and repellant activities against S. littoralis fifth instar larvae when applied to lettuce leaf disks at 0.01 and 0.1 UG/cm2, respectively; reduces the number of citric acid-induced coughs in guinea pigs at 100 mg/kg; decreases BALF neutrophil and macrophage infiltration and reduces peribronchial and perivascular inflammatory cell infiltration in a cigarette smoke-induced mouse model of acute lung inflammation from 1-10 mg/kg