Learn More
AVANTOR MASTERFLEX (VWR LLC) Conical-Bottom Tubes, 15 mL/PP/Plug; 25x20 Bags
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX1740026
Encompass
Corning tubes are available with either plug seal or flat top caps. Fifteen- and 50-mL tubes feature black printed graduations and a white writing patch; 250- and 500-mL tubes feature molded-in graduations. All are sterile and certified nonpyrogenic. The 50-mL tube 17400-44 is self-standing.
Catalog No. NC1870964