Learn More
LAKE CHARLES MFG 16x100mm Test Tubes, Polystyrene, Round BTM, 14ml Vol, Clear, Karter Scientific 206S5 (Case of 1000)
Catalog No. NC1507579
Encompass
Cylindrical test tube, manufactured from high quality polystyrene. PS Tubes are not autoclavable or dishwasher safe but are chemically resistant and clear like glass. Polystyrene has the properties of glass but at a much lower price point. Since polystyrene is not as hard as glass it will have minor scratches. Colors is clear
Catalog No. NC1507579 Supplier LAKE CHARLES MFG Supplier No. 206S5
Cylindrical test tube, manufactured from high quality polystyrene. PS Tubes are not autoclavable or dishwasher safe but are chemically resistant and clear like glass. Polystyrene has the properties of glass but at a much lower price point. Since polystyrene is not as hard as glass it will have minor scratches. Colors is clear