Learn More
Andwin Scientific TUBES 18ML 16X125MM 1000CS
SDP
Catalog No. NC0503962
Encompass_Preferred
The Culture Tubes feature resistance against chemical attack and weathering. In order to remove contamination, increase its strength, and lessen the chance of breakage, each tube is heat-treated at temperatures exceeding 500C (932F) immediately bef
Catalog No. NC0503962 Supplier Andwin Scientific Supplier No. 47729578
The Culture Tubes feature resistance against chemical attack and weathering. In order to remove contamination, increase its strength, and lessen the chance of breakage, each tube is heat-treated at temperatures exceeding 500C (932F) immediately bef