Learn More
Beckman Coulter TUBES 5/8X2-3/8 BELL OPTISEAL
Catalog No. NC9732446
Encompass_Preferred
OptiSeal Polyallomer Tubes,Quantity:56,Size(mm):16 x 60, Vol.(mL): 8.9
Catalog No. NC9732446 Supplier Beckman Coulter Supplier No. 361623
OptiSeal Polyallomer Tubes,Quantity:56,Size(mm):16 x 60, Vol.(mL): 8.9