Learn More
Med Vet International Dynarex Endotracheal Tubes - Uncuffed, 5.5 mm, 10/Box
Catalog No. 502831076
Encompass_Preferred
Dynarex Resp-O2™ Endotracheal Tubes are designed from premium plastic materials to efficiently deliver oxygen at higher concentrations. They may also be used to administer medications directly into the lungs. These Endotracheal Tubes are contoured for improved ease of administration and feature smoothly formed Murphy eye holes for improved comfort. Dynarex Endotracheal Tubes are available cuffed or uncuffed, and with or without an intubating stylette to help support the tube and guide it through the patient’s airway. Dynarex Resp-O2™ Endotracheal Tubes are high-quality plastic tubes designed to safely and effectively deliver higher concentrations of oxygen and medications directly into patients’ lungs.
Catalog No. 50-283-1076 Supplier Med Vet International Supplier No. 36227
Dynarex Resp-O2™ Endotracheal Tubes are designed from premium plastic materials to efficiently deliver oxygen at higher concentrations. They may also be used to administer medications directly into the lungs. These Endotracheal Tubes are contoured for improved ease of administration and feature smoothly formed Murphy eye holes for improved comfort. Dynarex Endotracheal Tubes are available cuffed or uncuffed, and with or without an intubating stylette to help support the tube and guide it through the patient’s airway. Dynarex Resp-O2™ Endotracheal Tubes are high-quality plastic tubes designed to safely and effectively deliver higher concentrations of oxygen and medications directly into patients’ lungs.