Learn More
Beckman Coulter TUBES PP 1 X 3-1/2 INCHES (BX-50)
Catalog No. NC9530407
Encompass_Preferred
Polypropylene Tube, Open-Top, Thinwall, 38.5 mL, 25 x 89mm, Quantity: 50Pk
Catalog No. NC9530407 Supplier Beckman Coulter Supplier No. 326823
Polypropylene Tube, Open-Top, Thinwall, 38.5 mL, 25 x 89mm, Quantity: 50Pk