Learn More
Beckman Coulter TUBES PA 5/8 X 4 (BOX-50)
Catalog No. NC9605996
Encompass_Preferred
Polyallomer Tubes,Quantity:50,Size(mm):16 x 96, Vol.(mL): 16.5
Catalog No. NC9605996 Supplier Beckman Coulter Supplier No. 337986
Polyallomer Tubes,Quantity:50,Size(mm):16 x 96, Vol.(mL): 16.5