Learn More
Beckman Coulter TUBES PP 9/16 X 3-1/2 (BOX-50)
Catalog No. NC9336079
Encompass_Preferred
Polypropylene Tubes,Thinwall, 13.2 mL, 14 x 89mm, Quantity: 50Pk
Catalog No. NC9336079 Supplier Beckman Coulter Supplier No. 331372
Polypropylene Tubes,Thinwall, 13.2 mL, 14 x 89mm, Quantity: 50Pk