Learn More
Beckman Coulter 3.5 mL, Open-Top Thickwall Polypropylene Tube, 13 x 51mm, 25/pk
Catalog No. NC9529688
Encompass_Preferred
Tubes compatible with rotor TLA-100.3 (p/n 349481) Polyallomer 13x51mm 3.5mL
Catalog No. NC9529688 Supplier Beckman Coulter Supplier No. 349623
Tubes compatible with rotor TLA-100.3 (p/n 349481) Polyallomer 13x51mm 3.5mL