Learn More
Beckman Coulter TUBES PA TK WALL 7X20MM (100)
Catalog No. NC9668582
Encompass_Preferred
Thickwall Polyallomer Tubes,Quantity:100,Size(mm):7 x 20, Vol.(microliter): 230
Catalog No. NC9668582 Supplier Beckman Coulter Supplier No. 343621
Thickwall Polyallomer Tubes,Quantity:100,Size(mm):7 x 20, Vol.(microliter): 230