Learn More
Beckman Coulter TUBES PC 1 X 3-1/2 (BOX-25)
Catalog No. NC9421069
Encompass_Preferred
Thickwall Polycarbonate Tubes,Quantity:25,Size(mm):25 x 89, Vol.(mL): 30.0
Catalog No. NC9421069 Supplier Beckman Coulter Supplier No. 355631
Thickwall Polycarbonate Tubes,Quantity:25,Size(mm):25 x 89, Vol.(mL): 30.0