Learn More
Beckman Coulter TUBES PC TK WALL 8X34MM (BOX-100)
Catalog No. NC9425062
Encompass_Preferred
Thickwall Tube, Polycarbonate, 0.5mL, 8x34mm, Quantity: 100/Box
Catalog No. NC9425062 Supplier Beckman Coulter Supplier No. 343776
Thickwall Tube, Polycarbonate, 0.5mL, 8x34mm, Quantity: 100/Box