Learn More
Mitegen Tubes, Wizard Precipitant Synergy, 192 formulations, 10 mL volumes
SDP
Catalog No. 501492099
Encompass_Preferred
Discover more protein crystallization hits with this unique screen by employing synergistic effects of mechanistically distinct precipitating agents (salts, organic solvents, polyethylene glycols, additives, pH). The Wizard Precipitant Synergy crystallization consists of 192 formulations, to be used for initial protein crystallization screening and crystallization optimization. The application of this screen is advised in particular for protein targets that have failed to yield hits with traditional random sparse matrix crystallization approaches.
Catalog No. 50-149-2099 Supplier Mitegen Supplier No. 1009539
Discover more protein crystallization hits with this unique screen by employing synergistic effects of mechanistically distinct precipitating agents (salts, organic solvents, polyethylene glycols, additives, pH). The Wizard Precipitant Synergy crystallization consists of 192 formulations, to be used for initial protein crystallization screening and crystallization optimization. The application of this screen is advised in particular for protein targets that have failed to yield hits with traditional random sparse matrix crystallization approaches.