Learn More
Beckman Coulter TUBES QS PA 13 X 32MM (BOX-50
Catalog No. NC9521284
Encompass_Preferred
Tubes compatible with rotor TLA-100.3 (p/n 349481) Polyallomer 13x32mm 3.5mL
Catalog No. NC9521284 Supplier Beckman Coulter Supplier No. 349621
Tubes compatible with rotor TLA-100.3 (p/n 349481) Polyallomer 13x32mm 3.5mL