Learn More
Beckman Coulter TUBES QS PP 5/8 X 3 (BOX-50)
Catalog No. NC9195096
Encompass_Preferred
Quick-Seal Standard Tube, Polypropylene, 13.5mL, 16x76mm, Quantity: 50/Box
Catalog No. NC9195096 Supplier Beckman Coulter Supplier No. 342413
Quick-Seal Standard Tube, Polypropylene, 13.5mL, 16x76mm, Quantity: 50/Box