Learn More
Beckman Coulter 4.2 mL Quick-Seal Bell-Top Polypropylene Tube, 16 x 38mm - 50Pk
Catalog No. NC0943795
Encompass_Preferred
Quick-Seal Polypropylene Tube
For use in High Performance and High Capacity Rotors: JS-24.15 (Part Number 362396); Required Spacer: 355579
For use in Ultracentrifuge Rotors: MLA-55
Catalog No. NC0943795 Supplier Beckman Coulter Supplier No. 356562
Quick-Seal Polypropylene Tube
For use in High Performance and High Capacity Rotors: JS-24.15 (Part Number 362396); Required Spacer: 355579
For use in Ultracentrifuge Rotors: MLA-55