Learn More
Andwin Scientific TUBES STRIP W/CAPS 125/PKTUBE
SDP
Catalog No. NC9883941
Encompass_Preferred
The PCR 8-Well Tube Strips are recommended for use for all PCR protocols. This product can be used with most popular thermal cycler blocks. The 0.2 mL tubes feature smooth, thin, uniform walls that allow accurate thermal transfer. Caps that fit preci
Catalog No. NC9883941 Supplier Andwin Scientific Supplier No. 20170004
The PCR 8-Well Tube Strips are recommended for use for all PCR protocols. This product can be used with most popular thermal cycler blocks. The 0.2 mL tubes feature smooth, thin, uniform walls that allow accurate thermal transfer. Caps that fit preci